1-溴-2,3,5-三氟苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,3,5-Trifluorobromobenzene

B2970-1G B2970-5G
TCIAB2970-1GEA 0 CNY
TCIAB2970-1G TCIAB2970-5G
1-溴-2,3,5-三氟苯 ≥98.0% (by GC)
1-溴-2,3,5-三氟苯
分子式: C₆H₂BrF₃
分子量: 210.98 g/mol
闪点: 32°C
MDL编号: MFCD00012232
CAS号: 133739-70-5
UN号: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05550599e4afb3906a4d8a0f6966d9ff25e72d1a&vid=05550599e4afb3906a4d8a0f6966d9ff25e72d1a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR