4-硝基-3-三氟甲基苯胺 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-硝基-α,α,α-三氟间甲苯胺, 5-氨基-2-硝基三氟甲苯

A0377-25G A0377-5G
TCIAA0377-25GEA 0 CNY
TCIAA0377-25G TCIAA0377-5G
4-硝基-3-三氟甲基苯胺 ≥98.0% (by GC)
4-硝基-3-三氟甲基苯胺
分子式: C₇H₅F₃N₂O₂
分子量: 206.12 g/mol
溶点: 129
MDL编号: MFCD00014717
CAS号: 393-11-3
EINECS: 206-884-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70144ed2aa1fd76a0e7a68b2590bfa24c9c41526&vid=70144ed2aa1fd76a0e7a68b2590bfa24c9c41526&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Orange to Brown to Dark red powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR