4-氯-2-(三氟甲基)苯胺 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2-氨基-5-氯三氟甲苯, 4-氯-α,α,α-三氟邻甲苯胺

A1058-500G A1058-25G
TCIAA1058-500GEA 0 CNY
TCIAA1058-500G TCIAA1058-25G
4-氯-2-(三氟甲基)苯胺 ≥98.0% (by GC)
4-氯-2-(三氟甲基)苯胺
分子式: C₇H₅ClF₃N
分子量: 195.57 g/mol
沸点: 77°C
溶点: 9
闪点: 95°C
存储条件: Refrigerator
MDL编号: MFCD00007841
CAS号: 445-03-4
EINECS: 207-151-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9edb4c0a2eb68d9d0056b7e91c3c3c47a1f85a0d&vid=9edb4c0a2eb68d9d0056b7e91c3c3c47a1f85a0d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR