4-(三氟甲氧基)苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: α,α,α-三氟对茴香胺

T1345-5G T1345-25G
TCIAT1345-5GEA 0 CNY
TCIAT1345-5G TCIAT1345-25G
4-(三氟甲氧基)苯胺 ≥98.0% (by GC, titration analysis)
4-(三氟甲氧基)苯胺
分子式: C₇H₆F₃NO
分子量: 177.13 g/mol
沸点: 83°C
密度: 1.31
闪点: 81°C
MDL编号: MFCD00041314
CAS号: 461-82-5
EINECS: 207-317-5
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72cc4c86ac8f99162d8ce4ca0161c546d249be73&vid=72cc4c86ac8f99162d8ce4ca0161c546d249be73&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR