5-Bromo-2-fluoroaniline ≥95.0% (by GC, titration analysis)

供应商: TCI
Warning

B3043-25G B3043-5G
TCIAB3043-25GEA 0 CNY
TCIAB3043-25G TCIAB3043-5G
5-Bromo-2-fluoroaniline ≥95.0% (by GC, titration analysis)
5-Bromo-2-fluoroaniline
分子式: C₆H₅BrFN
分子量: 190.01 g/mol
沸点: 105°C
MDL编号: MFCD03094181
CAS号: 2924-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb194b802474819c52235f4b15085136fa6ead98&vid=eb194b802474819c52235f4b15085136fa6ead98&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR