3,5-二(三氟甲基)苯胺 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
5-氨基-α,α,α,α′,α′,α′-六氟间二甲苯, α,α,α,α′,α′,α′-六氟-3,5-二甲基苯胺
B1324-10G
B1324-25G
B1324-250G
TCIAB1324-10GEA
0
CNY
TCIAB1324-10G
TCIAB1324-25G
TCIAB1324-250G
3,5-二(三氟甲基)苯胺 ≥97.0% (by GC)
3,5-二(三氟甲基)苯胺
分子式:
C₈H₅F₆N 沸点: 85°C 溶点: 3 °C 密度: 1,4753 g/cm³ (20 °C) 闪点: 98°C 存储条件: Ambient |
MDL编号:
MFCD00000394 CAS号: 328-74-5 EINECS: 206-335-0 ADR: 6.1,III |
|
产品技术规格
Appearance | Colorless to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...