2-氟-5-甲基苯胺 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
6-氟间甲苯胺
F0528-25G
F0528-5G
TCIAF0528-25GEA
0
CNY
TCIAF0528-25G
TCIAF0528-5G
2-氟-5-甲基苯胺 ≥98.0% (by GC, titration analysis)
2-氟-5-甲基苯胺
分子式:
C₇H₈FN 分子量: 125.15 g/mol 沸点: 90°C 闪点: 82°C |
MDL编号:
MFCD00007654 CAS号: 452-84-6 |
|
产品技术规格
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...