2-溴-4-氟苯胺 ≥98.0% (by GC)

供应商: TCI
Warning

B1829-25G B1829-5G
TCIAB1829-25GEA 0 CNY
TCIAB1829-25G TCIAB1829-5G
2-溴-4-氟苯胺 ≥98.0% (by GC)
2-溴-4-氟苯胺
分子式: C₆H₅BrFN
分子量: 190.01 g/mol
沸点: 221°C
溶点: 41
MDL编号: MFCD00042462
CAS号: 1003-98-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a997892c616da8a790a8fa852262314b38408423&vid=a997892c616da8a790a8fa852262314b38408423&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Brown powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR