2,5-二(三氟甲基)苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,5-Bis(trifluoromethyl)aniline
B2544-25G
B2544-5G
TCIAB2544-25GEA
0
CNY
TCIAB2544-25G
TCIAB2544-5G
2,5-二(三氟甲基)苯胺 ≥98.0% (by GC)
2,5-二(三氟甲基)苯胺
分子式:
C₈H₅F₆N 分子量: 229.12 g/mol 沸点: 177°C 闪点: 71°C |
MDL编号:
MFCD00074940 CAS号: 328-93-8 |
|
产品技术规格
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...