N-甲基-间甲苯胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: N-甲基-3-甲苯胺

M0446-25ML M0446-500ML
TCIAM0446-25MLEA 0 CNY
TCIAM0446-25ML TCIAM0446-500ML
N-甲基-间甲苯胺 ≥98.0% (by GC, titration analysis)
N-甲基-间甲苯胺
分子式: C₈H₁₁N
沸点: 207°C
密度: 0,9467 g/cm³ (15 °C)
闪点: 100°C
MDL编号: MFCD00035787
CAS号: 696-44-6
EINECS: 211-795-0
UN号: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06cd08d86824ad743782ff2d5acfb03e907b2e11&vid=06cd08d86824ad743782ff2d5acfb03e907b2e11&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR