N,N-二甲基间甲苯胺 ≥97.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: N,N,3-Trimethylaniline, 3-(Dimethylamino)toluene, N,N-Dimethyl-3-methylaniline, 3,N,N-Trimethylaniline

D0806-25ML D0806-500ML
TCIAD0806-25MLEA 0 CNY
TCIAD0806-25ML TCIAD0806-500ML
N,N-二甲基间甲苯胺 ≥97.0% (by GC, titration analysis)
N,N-二甲基间甲苯胺
分子式: C₉H₁₃N
分子量: 135.21 g/mol
沸点: 91°C
溶点: -15
闪点: 85°C
MDL编号: MFCD00008305
CAS号: 121-72-2
EINECS: 204-495-6
UN号: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78b1ce54b4d339154aa7c8f4f00e64e831c11b46&vid=78b1ce54b4d339154aa7c8f4f00e64e831c11b46&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR