2,6-二氟苄基氯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene

D2337-25G D2337-5G
TCIAD2337-25GEA 0 CNY
TCIAD2337-25G TCIAD2337-5G
2,6-二氟苄基氯 ≥98.0% (by GC)
2,6-二氟苄基氯
分子式: C₇H₅ClF₂
沸点: 172 - 173 °C (640 -rr)
溶点: 32 °C
MDL编号: MFCD00191346
CAS号: 697-73-4
UN号: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67278f893714346e17430c62b41ba9e53094b8e7&vid=67278f893714346e17430c62b41ba9e53094b8e7&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR