2,5-二氯溴苄 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,5-Dichlorobenzyl bromide, 2-(Bromomethyl)-1,4-dichlorobenzene

D3548-25G D3548-5G
TCIAD3548-25GEA 0 CNY
TCIAD3548-25G TCIAD3548-5G
2,5-二氯溴苄 ≥98.0% (by GC)
2,5-二氯溴苄
分子式: C₇H₅BrCl₂
沸点: 138°C
溶点: 39…41 °C
MDL编号: MFCD00236034
CAS号: 85482-13-9
UN号: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ffd61f75680f15896e9a609929880adccd59bbd&vid=3ffd61f75680f15896e9a609929880adccd59bbd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR