α-Bromo-2,3-dichlorotoluene ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
alpha-Bromo-2,3-dichlorotoluene, 2,3-Dichlorobenzyl bromide, 1-(Bromomethyl)-2,3-dichlorobenzene
D2920-25G
D2920-5G
TCIAD2920-25GEA
0
CNY
TCIAD2920-25G
TCIAD2920-5G
α-Bromo-2,3-dichlorotoluene ≥98.0% (by GC, titration analysis)
α-Bromo-2,3-dichlorotoluene
分子式:
C₇H₅BrCl₂ 分子量: 239.93 g/mol 沸点: 144°C |
MDL编号:
MFCD03701317 CAS号: 57915-78-3 UN号: 3261 ADR: 8 II |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...