3-甲氧基苄胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole

M0851-25ML M0851-5ML
TCIAM0851-25MLEA 0 CNY
TCIAM0851-25ML TCIAM0851-5ML
3-甲氧基苄胺 ≥98.0% (by GC, titration analysis)
3-甲氧基苄胺
分子式: C₈H₁₁NO
沸点: 141°C
密度: 1,07 g/cm³ (20 °C)
MDL编号: MFCD00008115
CAS号: 5071-96-5
EINECS: 225-779-6
UN号: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b382fb111a6e70b7b5fdc93a64021d4531a83de5&vid=b382fb111a6e70b7b5fdc93a64021d4531a83de5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR