蒲桃酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 庚二酸

P0435-25G P0435-500G
TCIAP0435-25GEA 0 CNY
TCIAP0435-25G TCIAP0435-500G
蒲桃酸 ≥98.0% (by GC, titration analysis)
蒲桃酸
分子式: C₇H₁₂O₄
沸点: 215°C
溶点: 101…103 °C
密度: 1,329 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00004425
CAS号: 111-16-0
EINECS: 203-840-8
Merck Index: 14,07431
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=182acfafee2c12421e269cc24c268a469d20adf8&vid=182acfafee2c12421e269cc24c268a469d20adf8&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR