己二酸二乙酯 ≥99.0% (by GC)
供应商: TCI
Synonyms:
NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363
A0162-500ML
A0162-25ML
TCIAA0162-500MLEA
0
CNY
TCIAA0162-500ML
TCIAA0162-25ML
己二酸二乙酯 ≥99.0% (by GC)
己二酸二乙酯
分子式:
C₁₀H₁₈O₄ 分子量: 202.25 g/mol 沸点: 251 溶点: -19.8 闪点: 127°C 存储条件: Ambient |
MDL编号:
MFCD00009215 CAS号: 141-28-6 EINECS: 205-477-0 Merck Index: 14,00162 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Acid Value | max. 0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...