叔丁基氢醌 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ
B0833-25G
B0833-500G
TCIAB0833-25GEA
0
CNY
TCIAB0833-25G
TCIAB0833-500G
叔丁基氢醌 ≥98.0% (by GC)
叔丁基氢醌
分子式:
C₁₀H₁₄O₂ 分子量: 166.22 g/mol 沸点: 273 溶点: 128 闪点: 171 存储条件: Ambient |
MDL编号:
MFCD00002344 CAS号: 1948-33-0 EINECS: 217-752-2 UN号: 3077 ADR: 9 III |
|
产品技术规格
Appearance | White to Light yellow to Light red powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 127.0 to 131.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...