2,5-二氟苯基异氰酸酯 ≥98.0% (通过GC)

供应商: TCI
Danger

D3695-1G D3695-5G
TCIAD3695-1GEA 0 CNY
TCIAD3695-1G TCIAD3695-5G
2,5-二氟苯基异氰酸酯 ≥98.0% (通过GC)
2,5-二氟苯基异氰酸酯
分子式: C₇H₃F₂NO
沸点: 54°C
密度: 1,32 g/cm³ (20 °C)
闪点: 56°C
存储条件: Refrigerator
MDL编号: MFCD00013843
CAS号: 39718-32-6
UN号: 3080
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3e244e7ad60f1d3996922d23fe55bc01687735c&vid=c3e244e7ad60f1d3996922d23fe55bc01687735c&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR