丙二酸二甲酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
1,3-Propanedioic acid dimethyl ester, Malonic acid dimethyl ester
M0030-25G
M0030-500G
TCIAM0030-25GEA
0
CNY
TCIAM0030-25G
TCIAM0030-500G
丙二酸二甲酯 ≥98.0% (by GC)
丙二酸二甲酯
分子式:
C₅H₈O₄ 沸点: 184°C 溶点: –62 °C 密度: 1,15 g/cm³ (20 °C) 闪点: 91°C 存储条件: Ambient |
MDL编号:
MFCD00008460 CAS号: 108-59-8 EINECS: 203-597-8 Merck Index: 14,06096 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...