1,3-二甲基-2-碘苯 ≥98.0% (通过GC)

供应商: TCI
Warning

I0752-25G I0752-5G
TCIAI0752-25GEA 0 CNY
TCIAI0752-25G TCIAI0752-5G
1,3-二甲基-2-碘苯 ≥98.0% (通过GC)
1,3-二甲基-2-碘苯
分子式: C₈H₉I
沸点: 230°C
溶点: 11°C
密度: 1,6518 g/cm³ (20 °C)
闪点: 102°C
MDL编号: MFCD00019012
CAS号: 608-28-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85352959cb55e2b493efe6555169acbb74077bf3&vid=85352959cb55e2b493efe6555169acbb74077bf3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR