3,5-二甲氧基甲苯 ≥97.0% (通过GC)

供应商: TCI
Warning

D2526-25G D2526-5G
TCIAD2526-25GEA 0 CNY
TCIAD2526-25G TCIAD2526-5G
3,5-二甲氧基甲苯 ≥97.0% (通过GC)
3,5-二甲氧基甲苯
分子式: C₉H₁₂O₂
沸点: 112°C
闪点: 101°C
MDL编号: MFCD00015435
CAS号: 4179-19-5
EINECS: 224-048-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c9ca1ff54cd9e76c9fb6aaa1a0e675ec8c34322&vid=7c9ca1ff54cd9e76c9fb6aaa1a0e675ec8c34322&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR