2′,6′-二氟苯乙酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone

D3559-25G D3559-5G
TCIAD3559-25GEA 0 CNY
TCIAD3559-25G TCIAD3559-5G
2′,6′-二氟苯乙酮 ≥98.0% (by GC)
2′,6′-二氟苯乙酮
分子式: C₈H₆F₂O
沸点: 79°C
密度: 1,19 g/cm³ (20 °C)
闪点: 76°C
MDL编号: MFCD00000328
CAS号: 13670-99-0
EINECS: 237-151-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87f2b38cd8d6be85926fd42e19859c5c435a9bce&vid=87f2b38cd8d6be85926fd42e19859c5c435a9bce&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR