2,6-Difluorobenzyl alcohol ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(2,6-Difluorophenyl)methanol
D2507-25G
D2507-5G
TCIAD2507-25GEA
0
CNY
TCIAD2507-25G
TCIAD2507-5G
2,6-Difluorobenzyl alcohol ≥97.0% (by GC)
2,6-Difluorobenzyl alcohol
分子式:
C₇H₆F₂O 沸点: 88°C 密度: 1,3 g/cm³ (20 °C) 闪点: 88°C |
MDL编号:
MFCD00004603 CAS号: 19064-18-7 EINECS: 242-792-2 |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...