3,5-二氯苯甲醇 ≥98.0% (by GC)
供应商: TCI
D3791-25G
D3791-5G
TCIAD3791-25GEA
0
CNY
TCIAD3791-25G
TCIAD3791-5G
3,5-二氯苯甲醇 ≥98.0% (by GC)
3,5-二氯苯甲醇
分子式:
C₇H₆Cl₂O 分子量: 177.03 g/mol 沸点: 264 溶点: 82 |
MDL编号:
MFCD00004634 CAS号: 60211-57-6 EINECS: 262-109-1 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...