2′,4′-二氯苯乙酮 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2′,4′-Dichloroacetophenone
D1490-25G
D1490-500G
TCIAD1490-25GEA
0
CNY
TCIAD1490-25G
TCIAD1490-500G
2′,4′-二氯苯乙酮 ≥98.0% (by GC)
2′,4′-二氯苯乙酮
分子式:
C₈H₆Cl₂O 分子量: 189.04 g/mol 沸点: 247 溶点: 42 |
MDL编号:
MFCD00000581 CAS号: 2234-16-4 EINECS: 218-780-8 |
|
产品技术规格
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...