2′,6′-二氯苯乙酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-(2,6-Dichlorophenyl)ethanone

D4119-1G D4119-5G
TCIAD4119-1GEA 0 CNY
TCIAD4119-1G TCIAD4119-5G
2′,6′-二氯苯乙酮 ≥98.0% (by GC)
2′,6′-二氯苯乙酮
分子式: C₈H₆Cl₂O
分子量: 189.04 g/mol
沸点: 141°C
MDL编号: MFCD00016339
CAS号: 2040-05-3
EINECS: 218-035-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbeed74d1d2026853771b7875be8e0e2064373f3&vid=fbeed74d1d2026853771b7875be8e0e2064373f3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR