2,4-Dibromotoluene ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 2,4-Dibromo-1-methylbenzene

D3591-1G D3591-5G
TCIAD3591-1GEA 0 CNY
TCIAD3591-1G TCIAD3591-5G
2,4-Dibromotoluene ≥98.0% (by GC)
2,4-Dibromotoluene
分子式: C₇H₆Br₂
沸点: 243°C
溶点: -10°C
密度: 1,85
MDL编号: MFCD00052985
CAS号: 31543-75-6
EINECS: 250-689-9

现在订购

产品技术规格

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR