四氟邻苯二腈 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 3,4,5,6-四氟邻苯二甲腈, 3,4,5,6-四氟邻苯二腈

T1402-25G T1402-5G
TCIAT1402-25GEA 0 CNY
TCIAT1402-25G TCIAT1402-5G
四氟邻苯二腈 ≥98.0% (by GC)
四氟邻苯二腈
分子式: C₈F₄N₂
分子量: 200.1 g/mol
MDL编号: MFCD00001774
CAS号: 1835-65-0
EINECS: 217-400-8
UN号: 3439
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5bed3cb4d8d0f52ee5db900fe9edc687eda8746&vid=b5bed3cb4d8d0f52ee5db900fe9edc687eda8746&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR