十二烷二酸 ≥99.0% (by GC)

供应商: TCI
Warning

Synonyms: 癸烷-1,10-二羧酸

D0013-25G D0013-500G
TCIAD0013-25GEA 0 CNY
TCIAD0013-25G TCIAD0013-500G
十二烷二酸 ≥99.0% (by GC)
十二烷二酸
分子式: C₁₂H₂₂O₄
分子量: 230.3 g/mol
沸点: 294
闪点: 220
存储条件: Freezer
MDL编号: MFCD00002735
CAS号: 693-23-2
EINECS: 211-746-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8628028465cd4580e57b78121afe3b1053edf93f&vid=8628028465cd4580e57b78121afe3b1053edf93f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 128.0 to 132.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR