(S)-(+)-α-甲氧基苯乙酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: O-甲基-L-扁桃酸

M0829-1G M0829-5G
TCIAM0829-1GEA 0 CNY
TCIAM0829-1G TCIAM0829-5G
(S)-(+)-α-甲氧基苯乙酸 ≥98.0% (by GC, titration analysis)
(S)-(+)-α-甲氧基苯乙酸
分子式: C₉H₁₀O₃
沸点: 130°C
溶点: 71…73 °C
MDL编号: MFCD00064216
CAS号: 26164-26-1
EINECS: 247-492-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b65eb04dfcd2d155ed75a6d3861ac93f52e86179&vid=b65eb04dfcd2d155ed75a6d3861ac93f52e86179&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Purity (Neutralization titration) min. 98.0 %
Melting point 64.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR