(S)-(+)-2-辛醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
D(+)-2-Octanol, (S)-Octan-2-ol, (S)-(+)-Hexyl methyl carbinol, (S)-(+)-2-Hydroxy octane, (S)-(+)-Methyl hexyl carbinol
O0144-25ML
O0144-5ML
TCIAO0144-25MLEA
0
CNY
TCIAO0144-25ML
TCIAO0144-5ML
(S)-(+)-2-辛醇 ≥98.0% (by GC)
(S)-(+)-2-辛醇
分子式:
C₈H₁₈O 分子量: 130.23 g/mol 沸点: 175 闪点: 76°C 存储条件: Ambient |
MDL编号:
MFCD00064283 CAS号: 6169-06-8 EINECS: 228-213-6 Merck Index: 14,06752 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | +9.0 to +11.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...