L(+)-Norleucine ≥99.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: H-Nle-OH, H-NorLeu-OH, (S)-2-Aminohexanoic acid, (S)-(+)-Norleucine

N0303-1G N0303-100MG
TCIAN0303-1GEA 0 CNY
TCIAN0303-1G TCIAN0303-100MG
L(+)-Norleucine ≥99.0% (by titrimetric analysis)
L(+)-Norleucine
分子式: C₆H₁₃NO₂
分子量: 131.17 g/mol
沸点: 246
溶点: 113
MDL编号: MFCD00064423
CAS号: 327-57-1
EINECS: 206-321-4
Merck Index: 14,06706
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc6eb94e0027712f4bbb4484f6891cfb7bd39f07&vid=cc6eb94e0027712f4bbb4484f6891cfb7bd39f07&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +22.0 to +24.0 deg (C=5, 5mol/L HCl)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR