(4-Chlorophenyl)acetic acid methyl ester ≥98.0% (by GC)

供应商: TCI

Synonyms: (4-Chlorophenyl)acetic acid methyll ester, Methyl (4-chlorophenyl)acetate, Methyl (4-Chlorophenyl)acetate, Methyl 4-chlorophenylacetate

M2039-25G M2039-5G
TCIAM2039-25GEA 0 CNY
TCIAM2039-25G TCIAM2039-5G
(4-Chlorophenyl)acetic acid methyl ester ≥98.0% (by GC)
(4-Chlorophenyl)acetic acid methyl ester
分子式: (C₆H₄Cl)CH₂COOCH₃
分子量: 184.62 g/mol
沸点: 115°C
MDL编号: MFCD00032743
CAS号: 52449-43-1
EINECS: 257-927-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21b940b3d82baa48b17cc6dc62e3ecdb02fbc299&vid=21b940b3d82baa48b17cc6dc62e3ecdb02fbc299&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR