3-苯基-1-丙醇 ≥98.0% (通过GC)

供应商: TCI
Warning

P0214-25ML P0214-500ML
TCIAP0214-25MLEA 0 CNY
TCIAP0214-25ML TCIAP0214-500ML
3-苯基-1-丙醇 ≥98.0% (通过GC)
3-苯基-1-丙醇
分子式: (C₆H₅)CH₂CH₂CH₂OH
沸点: 236°C
溶点: –18 °C
密度: 1,01 g/cm³ (20 °C)
闪点: 109°C
存储条件: Ambient
MDL编号: MFCD00002950
CAS号: 122-97-4
EINECS: 204-587-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbc638bb1163a465139d930b025fcaf144596b99&vid=cbc638bb1163a465139d930b025fcaf144596b99&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR