3,5-双(三氟甲基)苯乙酸 ≥97.0% (通过滴定分析)
供应商: TCI
|
Warning
|
B1976-1G
B1976-5G
TCIAB1976-1GEA
0
CNY
TCIAB1976-1G
TCIAB1976-5G
3,5-双(三氟甲基)苯乙酸 ≥97.0% (通过滴定分析)
3,5-双(三氟甲基)苯乙酸
分子式:
(C₆H₃(CF₃)₂)CH₂COOH 沸点: 276 °C (1013 hPa) 溶点: 121…125 °C |
MDL编号:
MFCD00009908 CAS号: 85068-33-3 EINECS: 285-295-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 125.0 to 129.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...