3,4-二氟苯乙酸 ≥97.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3,4-Difluorophenylacetic acid
D3771-1G
D3771-5G
TCIAD3771-1GEA
0
CNY
TCIAD3771-1G
TCIAD3771-5G
3,4-二氟苯乙酸 ≥97.0% (by GC, titration analysis)
3,4-二氟苯乙酸
分子式:
(C₆H₃F₂)CH₂COOH 分子量: 172.13 g/mol 沸点: 259 |
MDL编号:
MFCD00010002 CAS号: 658-93-5 EINECS: 211-527-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...