L-异亮氨酸 ≥98.0% (通过滴定分析), crystals

供应商: TCI

I0181-25G I0181-500G
TCIAI0181-25GEA 0 CNY
TCIAI0181-25G TCIAI0181-500G
L-异亮氨酸 ≥98.0% (通过滴定分析), crystals
L-异亮氨酸
分子式: H₃CCH₂CH(CH₃)CH(NH₂)COOH
溶点: 279…280 °C
存储条件: Ambient
MDL编号: MFCD00064222
CAS号: 73-32-5
EINECS: 200-798-2
Merck Index: 14,05179
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bceb3b99593bc0ea02f9ee279d1600fc4ad4e9b2&vid=948f14df5f68c9caf589185b29e22fc56e8d7842&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +38.0 to +43.0 deg (C=4, 6mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR