5-甲基喹喔啉 ≥98.0% (通过GC)

供应商: TCI
Warning

M1371-1G M1371-5G
TCIAM1371-1GEA 0 CNY
TCIAM1371-1G TCIAM1371-5G
5-甲基喹喔啉 ≥98.0% (通过GC)
5-甲基喹喔啉
分子式: C₉H₈N₂
沸点: 120°C
存储条件: Refrigerator
MDL编号: MFCD00012335
CAS号: 13708-12-8
EINECS: 237-246-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be3fe2ab8780b3021236db85f45fcdf859365027&vid=be3fe2ab8780b3021236db85f45fcdf859365027&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR