2-甲基间苯二酚 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 2,6-二羟基甲苯

M1109-25G M1109-500G
TCIAM1109-25GEA 0 CNY
TCIAM1109-25G TCIAM1109-500G
2-甲基间苯二酚 ≥98.0% (by GC)
2-甲基间苯二酚
分子式: C₇H₈O₂
沸点: 264 °C (1013 hPa)
溶点: 120…122 °C
闪点: 135 °C
存储条件: Ambient
MDL编号: MFCD00002271
CAS号: 608-25-3
EINECS: 210-155-8
UN号: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=782b0e21a8014b6a95d4d2526f4a8d0db7c9a9fa&vid=782b0e21a8014b6a95d4d2526f4a8d0db7c9a9fa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 123.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR