2-戊炔-1-醇 ≥98.0% (by GC)
供应商: TCI
Warning
P0815-25ML P0815-5ML
TCIAP0815-25MLEA 0 CNY
TCIAP0815-25ML TCIAP0815-5ML
2-戊炔-1-醇 ≥98.0% (by GC)
2-戊炔-1-醇
分子式: C₅H₈O
沸点: 154°C
密度: 0,913 g/cm³ (17 °C)
闪点: 62°C
存储条件: Ambient
MDL编号: MFCD00040915
CAS号: 6261-22-9
EINECS: 228-411-2
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cb3950054557fdae5aa0b4724aa10a6f3d0f385&vid=0cb3950054557fdae5aa0b4724aa10a6f3d0f385&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %