1-Octacosanol ≥93.0% (通过GC)

供应商: TCI

O0199-1G O0199-100MG
TCIAO0199-1GEA 0 CNY
TCIAO0199-1G TCIAO0199-100MG
1-Octacosanol ≥93.0% (通过GC)
1-Octacosanol
分子式: H₃C(CH₂)₂₆CH₂OH
沸点: 428 °C (1013 hPa)
MDL编号: MFCD00044770
CAS号: 557-61-9
EINECS: 209-181-2
Merck Index: 14,06744
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=966d07a52f1a47bf152e6154f3962c0fd7dcbf8d&vid=966d07a52f1a47bf152e6154f3962c0fd7dcbf8d&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR