1-氨基-2-丙醇 ≥92.0% (by GC)
供应商: TCI
Danger
Synonyms: (±)-1-氨基-2-丙醇, (±)-异丙醇胺
A0406-25G A0406-500G
TCIAA0406-25GEA 0 CNY
TCIAA0406-25G TCIAA0406-500G
1-氨基-2-丙醇 ≥92.0% (by GC)
1-氨基-2-丙醇
分子式: C₃H₉NO
分子量: 75.11 g/mol
沸点: 160°C
溶点: –2°C
闪点: 78°C
存储条件: Ambient
MDL编号: MFCD00008139
CAS号: 78-96-6
EINECS: 201-162-7
UN号: 2735
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116212e9f5407396698e573ee28392a7f3ea82d0&vid=116212e9f5407396698e573ee28392a7f3ea82d0&prid=web&clid=SAPDAM
现在订购
产品技术规格
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 92.0 %