2,6-二氟硝基苯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
2,6-Difluoronitrobenzene
D4123-1G
D4123-5G
TCIAD4123-1GEA
0
CNY
TCIAD4123-1G
TCIAD4123-5G
2,6-二氟硝基苯 ≥98.0% (by GC)
2,6-二氟硝基苯
分子式:
C₆H₃F₂NO₂ 沸点: 92°C |
MDL编号:
MFCD00192035 CAS号: 19064-24-5 EINECS: 242-793-8 |
|
产品技术规格
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...