2-(三氟甲氧基)苯胺 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
α,α,α-三氟邻茴香胺
T2145-25G
T2145-5G
TCIAT2145-25GEA
0
CNY
TCIAT2145-25G
TCIAT2145-5G
2-(三氟甲氧基)苯胺 ≥98.0% (by GC)
2-(三氟甲氧基)苯胺
分子式:
C₇H₆F₃NO 分子量: 177.13 g/mol 沸点: 63°C 闪点: 54°C |
MDL编号:
MFCD00035959 CAS号: 1535-75-7 EINECS: 216-257-9 UN号: 1993 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...