3,4'-二氟二苯甲酮 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(3,4-Difluorophenyl)(phenyl)methanone
D3576-25G
D3576-5G
TCIAD3576-25GEA
0
CNY
TCIAD3576-25G
TCIAD3576-5G
3,4'-二氟二苯甲酮 ≥98.0% (by GC)
3,4'-二氟二苯甲酮
分子式:
C₁₃H₈F₂O 分子量: 218.2 g/mol 沸点: 293 |
MDL编号:
MFCD00009892 CAS号: 85118-07-6 EINECS: 285-659-4 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...