3-(Trifluoromethoxy)benzonitrile ≥98.0% (by GC)
供应商: TCI
|
Danger
|
T2372-1G
T2372-5G
TCIAT2372-1GEA
0
CNY
TCIAT2372-1G
TCIAT2372-5G
3-(Trifluoromethoxy)benzonitrile ≥98.0% (by GC)
3-(Trifluoromethoxy)benzonitrile
分子式:
C₈H₄F₃NO 分子量: 187.12 g/mol 沸点: 236 溶点: 37 |
MDL编号:
MFCD00040959 CAS号: 52771-22-9 UN号: 3276 ADR: 6.1 III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...