4-氟联苯 ≥97.0% (by GC)
供应商: TCI
Synonyms:
PFB No. 3, 4-Fluoro-1,1-biphenyl
F0482-1G
F0482-5G
TCIAF0482-1GEA
0
CNY
TCIAF0482-1G
TCIAF0482-5G
4-氟联苯 ≥97.0% (by GC)
4-氟联苯
分子式:
C₁₂H₉F 分子量: 172.2 g/mol 沸点: 253°C 溶点: 75°C |
MDL编号:
MFCD00011650 CAS号: 324-74-3 EINECS: 206-304-1 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 77.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...