十一烷二酸 ≥97.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 壬烷二甲酸

N0333-25G N0333-5G
TCIAN0333-25GEA 0 CNY
TCIAN0333-25G TCIAN0333-5G
十一烷二酸 ≥97.0% (by GC, titration analysis)
十一烷二酸
分子式: C₁₁H₂₀O₄
沸点: 372 °C (1013 hPa)
溶点: 108…110 °C
密度: 1,195 g/cm³ (21 °C)
存储条件: Ambient
MDL编号: MFCD00004444
CAS号: 1852-04-6
EINECS: 217-440-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d8c93368efce65aa53182a7bfdfbc402f4a1fa&vid=f2d8c93368efce65aa53182a7bfdfbc402f4a1fa&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 109.0 to 113.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR