硫代乙醇酸 ≥98.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
巯基乙酸
M0052-25G
M0052-500G
TCIAM0052-25GEA
0
CNY
TCIAM0052-25G
TCIAM0052-500G
硫代乙醇酸 ≥98.0% (by titrimetric analysis)
硫代乙醇酸
分子式:
HSCH₂COOH 分子量: 92.12 g/mol 沸点: 96 溶点: -16.5 闪点: 119°C (closed cup) 存储条件: Refrigerator |
MDL编号:
MFCD00004876 CAS号: 68-11-1 EINECS: 200-677-4 UN号: 1940 ADR: 8 II Merck Index: 14,09336 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (Iodometric Titration) | min. 98.0 % |
Suitability for iron-analysis | Abs min 0.20 (near 535nm) in the presence of Fe (3ppm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...